missing translation for 'onlineSavingsMsg'
Learn More

Thioflavin T

Catalog No. p-200161
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC211760050 5 g Glass bottle
AC211760250 25 g Glass bottle
AC211761000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC211760050 Supplier Thermo Scientific Chemicals Supplier No. 211760050
Only null left

CAS: 2390-54-7 | C17H19ClN2S | 318.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2390-54-7
Molecular Formula C17H19ClN2S
Molecular Weight (g/mol) 318.86
MDL Number MFCD00011944
InChI Key JADVWWSKYZXRGX-UHFFFAOYSA-M
Synonym Basic Yellow 1, C.I. 49005
PubChem CID 16953
ChEBI CHEBI:76023
IUPAC Name 2-[4-(dimethylamino)phenyl]-3,6-dimethyl-1,3-benzothiazol-3-ium chloride
SMILES [Cl-].CN(C)C1=CC=C(C=C1)C1=[N+](C)C2=CC=C(C)C=C2S1

Specifications

Beilstein 27, 377
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most organic solvent
Infrared Spectrum Authentic
Formula Weight 318.85
Packaging Glass bottle
UV Spectrometry Complies
Color Ochre/Yellow to Yellow
Quantity 5 g
Physical Form Powder
Chemical Name or Material Thioflavin T
EINECSNumber 219-228-9
TSCA TSCA

RUO – Research Use Only