Learn More
Thiodiglycolic acid, 98%
CAS: 123-93-3 | C4H6O4S | 150.148 g/mol
Supplier: Thermo Scientific Chemicals A1788036
Description
It finds it applications in the production of [1,4]oxathiane-2,6-dione by heating. It is employed as an anti-oxidant and reducing agent in cosmetics industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds it applications in the production of [1,4]oxathiane-2,6-dione by heating. It is employed as an anti-oxidant and reducing agent in cosmetics industry.
Solubility
Solubility in water is almost transparent. (water, 4.00E+05 mg/L @ 25°C (exp); 1e+006 mg/L @ 25°C (est).
Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| Thiodiglycolic acid | |
| 123-93-3 | |
| MFCD00004359 | |
| 1764392 | |
| thiodiglycolic acid, 2,2'-thiodiacetic acid, thiodiacetic acid, mercaptodiacetic acid, tdga, dicarboxymethyl sulfide, thiodi acetic acid, carboxymethylthio acetic acid, 2,2'-thiodiethanoic acid, thiodiglycollic acid | |
| C(C(=O)O)SCC(=O)O | |
| 150.148 | |
| CHEBI:35012 | |
| 98% |
| 127°C to 131°C | |
| C4H6O4S | |
| UN3261 | |
| 149331 | |
| UVZICZIVKIMRNE-UHFFFAOYSA-N | |
| 2-(carboxymethylsulfanyl)acetic acid | |
| 31277 | |
| 150.15 | |
| 500 g |
Chemical Identifiers
| 123-93-3 | |
| 150.148 | |
| UVZICZIVKIMRNE-UHFFFAOYSA-N | |
| 31277 | |
| 2-(carboxymethylsulfanyl)acetic acid |
| C4H6O4S | |
| MFCD00004359 | |
| thiodiglycolic acid, 2,2'-thiodiacetic acid, thiodiacetic acid, mercaptodiacetic acid, tdga, dicarboxymethyl sulfide, thiodi acetic acid, carboxymethylthio acetic acid, 2,2'-thiodiethanoic acid, thiodiglycollic acid | |
| CHEBI:35012 | |
| C(C(=O)O)SCC(=O)O |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 204-663-9
RTECSNumber : AJ6475000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only