missing translation for 'onlineSavingsMsg'
Learn More

Thiocyanuric Acid 98.0+%, TCI America™

Catalog No. T093525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T093525G 25 g
T0935100G 100 g
T0935500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T093525G Supplier TCI America Supplier No. T093525G

Chemical Identifiers

CAS 638-16-4
Molecular Formula C3H3N3S3
Molecular Weight (g/mol) 177.258
MDL Number MFCD00006052
InChI Key WZRRRFSJFQTGGB-UHFFFAOYSA-N
Synonym trithiocyanuric acid, thiocyanuric acid, 1,3,5-triazine-2,4,6-trithiol, trismercaptotriazine, trimercaptocyanuric acid, cyanuric acid, trithio, usaf th-3, 1,3,5-triazine-2,4,6 1h,3h,5h-trithione, s-triazine-2,4,6-trithiol, 2,4,6-triazinetrithiol
PubChem CID 1268121
IUPAC Name 1,3,5-triazinane-2,4,6-trithione
SMILES C1(=S)NC(=S)NC(=S)N1

Specifications

Quantity 25 g
Formula Weight 177.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Thiocyanuric Acid
EINECSNumber (5)-1075
RTECSNumber XZ2830000
TSCA Yes