missing translation for 'onlineSavingsMsg'
Learn More

Thiocolchicoside 98.0+%, TCI America™

Catalog No. T291920MG
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T291920MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T291920MG Supplier TCI America Supplier No. T291920MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Synthetic colchicine derivative that acts as a selective GABAA receptor antagonist with muscle relaxant, anti-inflammatory, and analgesic properties.

Chemical Identifiers

CAS 602-41-5
Molecular Formula C27H33NO10S
Molecular Weight (g/mol) 563.618
MDL Number MFCD07995056
InChI Key LEQAKWQJCITZNK-AXHKHJLKSA-N
PubChem CID 9915886
IUPAC Name N-[(7S)-1,2-dimethoxy-10-methylsulfanyl-9-oxo-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide
SMILES CC(=O)NC1CCC2=CC(=C(C(=C2C3=CC=C(C(=O)C=C13)SC)OC)OC)OC4C(C(C(C(O4)CO)O)O)O

Specifications

Melting Point 220°C
Color Yellow
Quantity 20 mg
Formula Weight 563.62
Percent Purity ≥98.0% (HPLC)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material Thiocolchicoside
RTECSNumber AC2976015
TSCA No
Recommended Storage Refrigerator