Learn More
Thiocarbanilide, 98%, Thermo Scientific™
$48.94 - $200.87
Chemical Identifiers
| CAS | 102-08-9 |
|---|---|
| Molecular Formula | C13H12N2S |
| Molecular Weight (g/mol) | 228.31 |
| MDL Number | MFCD00004921 |
| InChI Key | FCSHMCFRCYZTRQ-UHFFFAOYSA-N |
| Synonym | thiocarbanilide, diphenylthiourea, 1,3-diphenyl-2-thiourea, n,n'-diphenylthiourea, sulfocarbanilide, sym-diphenylthiourea, thiourea, n,n'-diphenyl, stabilisator c, rhenocure ca, vulkacit ca |
| PubChem CID | 700999 |
| IUPAC Name | 1,3-diphenylthiourea |
| SMILES | S=C(NC1=CC=CC=C1)NC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC138701000
|
N/A | 100g | Plastic bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC138700010
|
N/A | 1kg | Plastic bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C13H12N2S | |
| MFCD00004921 | |
| thiocarbanilide, diphenylthiourea, 1,3-diphenyl-2-thiourea, n,n'-diphenylthiourea, sulfocarbanilide, sym-diphenylthiourea, thiourea, n,n'-diphenyl, stabilisator c, rhenocure ca, vulkacit ca | |
| 1,3-diphenylthiourea |
Specifications
| 102-08-9 | |
| Authentic | |
| C13H12N2S | |
| MFCD00004921 | |
| 15,3373 | |
| Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform | |
| S=C(NC1=CC=CC=C1)NC1=CC=CC=C1 | |
| 228.31 | |
| 228.32 | |
| Thiocarbanilide, 98% |
| 0.3% max. (60°C) | |
| 97.5% min. (HPLC) | |
| C6H5NHCSNHC6H5 | |
| 12,394 | |
| thiocarbanilide, diphenylthiourea, 1,3-diphenyl-2-thiourea, n,n'-diphenylthiourea, sulfocarbanilide, sym-diphenylthiourea, thiourea, n,n'-diphenyl, stabilisator c, rhenocure ca, vulkacit ca | |
| FCSHMCFRCYZTRQ-UHFFFAOYSA-N | |
| 1,3-diphenylthiourea | |
| 700999 | |
| 98% |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Suspected of damaging the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Warning
EINECSNumber : 203-004-2
RTECSNumber : FE1225000
TSCA : TSCA
RUO – Research Use Only