missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thioacetamide, For Chlorine, Chlorine Dioxide, 0.25% (w/v), LabChem™
Specifications
Specifications
| Color | Clear, colorless |
| Format | Liquid |
| Packaging | Poly Bottle |
| Quantity | 125 mL |
| Storage Requirements | Room Temperature |
| Type | Thioacetamide |