missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thioacetamide, For Chlorine, Chlorine Dioxide, 0.25% (w/v), LabChem™
Supplier: LabChem LC260101
Chemical Identifiers
| Clear, colorless | |
| Poly Bottle | |
| Thioacetamide |
| Liquid | |
| Room Temperature |
Specifications
| Clear, colorless | |
| Poly Bottle | |
| Room Temperature |
| Liquid | |
| 500 mL | |
| Thioacetamide |