Learn More
Epredia™ Thinline Slotted Cassettes in Tube Packs
For routine tissue samples, cassettes are compatible with all Epredia PrintMate™ Cassette Printers and have a front lid hinge with color-matched, attached lids available in 12 different color varieties.
$1581.88 - $1581.88
Specifications
| Closure Included | Yes |
|---|---|
| Quantity | 750/Cs. |
| Pore Style | Slots |
| Closure Type | Attached Lid, Front Hinge |
| PrintMate Compatible | Yes |
| Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
|
B851736BE
|
Epredia™
B851736BE |
Beige |
Case of 750 Each for $1,581.88
|
|
|||||
|
B851736BL
|
Epredia™
B851736BL |
Blue |
Case of 750 Each for $1,581.88
|
|
|||||
|
B851736GN
|
Epredia™
B851736GN |
Green |
Case of 750 Each for $1,581.88
|
|
|||||
|
B851736GY
|
Epredia™
B851736GY |
Gray |
Case of 750 Each for $1,581.88
|
|
|||||
|
B851736LA
|
Epredia™
B851736LA |
Lavender |
Case of 750 Each for $1,581.88
|
|
|||||
|
B851736LG
|
Epredia™
B851736LG |
Light Green |
Case of 750 Each for $1,581.88
|
|
|||||
|
B851736LI
|
Epredia™
B851736LI |
Lilac |
Case of 750 Each for $1,581.88
|
|
|||||
|
B851736OR
|
Epredia™
B851736OR |
Orange |
Case of 750 Each for $1,581.88
|
|
|||||
|
B851736TN
|
Epredia™
B851736TN |
Tan |
Case of 750 Each for $1,581.88
|
|
|||||
|
B851736WH
|
Epredia™
B851736WH |
White |
Case of 750 Each for $1,581.88
|
|
|||||
|
B851736YW
|
Epredia™
B851736YW |
Yellow |
Case of 750 Each for $1,581.88
|
|
|||||
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifications
| Yes | |
| Slots | |
| Yes | |
| Slotted Cassette |
| 750/Cs. | |
| Attached Lid, Front Hinge | |
| For Use With PrintMate™ Printers |