missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Thinline Slotted Cassettes in Tube Packs

Catalog No. B851736BE
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
B851736BE Beige
B851736BL Blue
B851736GN Green
B851736GY Gray
B851736LA Lavender
B851736LG Light Green
B851736LI Lilac
B851736OR Orange
B851736TN Tan
B851736WH White
B851736YW Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. B851736BE Supplier Epredia™ Supplier No. B851736BE

For routine tissue samples, cassettes are compatible with all Epredia PrintMate™ Cassette Printers and have a front lid hinge with color-matched, attached lids available in 12 different color varieties.

Printing

  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime

Grossing, Processing and Embedding

  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples

Sectioning

  • Precision engineering for solid connection to microtomes

Archiving

  • Cassette surface texture optimizes scannability for archiving

Specifications

Pore Style Slots
Quantity 750/Cs.
Closure Included Yes
Closure Type Attached Lid, Front Hinge
PrintMate Compatible Yes
Color Beige
For Use With (Equipment) For Use With PrintMate™ Printers
Product Type Slotted Cassette