missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One ThinCert™ 96 Well HTS Insert
Thincert™ 96 Well HTS inserts are indispensable tools for high-throughput screening and assays. Designed for precision, they enable researchers to elevate the quality and efficiency of their work.
Supplier: Greiner Bio-One 655641
Description
- Optimal tool for generating tissue models (e.g., endothelia and epithelia) for transport, uptake, or co-culture studies in submersed as well as air-lift culture with an automation-friendly format
- By combining high membrane permeability and best possible transparency, Greiner Bio-One is offering a unique 96 well insert version, exclusively designed for applications that require both high diffusion rates as well as microscopic readouts.
- Special pore arrangement and a high pore density of 2 x 107/cm2.
- Patented assembly technology: robust and reliable bonding of membrane
- Excellent membrane flatness: optimal and reproducible cell culture conditions
- Optimized fit: prevention of media wicking
- Enlarged access ports: convenient access for pipettes, robots and TEER electrodes
Chemical Identifiers
| No Barcode | |
| 8 x 12 | |
| Polycarbonate | |
| Sterile |
| Standard | |
| Clear | |
| 96 | |
| Flat Bottom (F-well) |
Specifications
| No Barcode | |
| 8 x 12 | |
| Insert with Transparent Membrane | |
| 0.15 to 1.7 μL | |
| 15 to 160 μL | |
| Tissue Culture |
| Standard | |
| Clear | |
| Polycarbonate | |
| 96 | |
| Sterile | |
| Flat Bottom (F-well) |