missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One ThinCert™ 96 Well HTS Insert

Catalog No. 01670254
Change view
Click to view available options
Product Type:
Insert with Permeable Membrane
Insert with Transparent Membrane
Receiver Plate
Surface Treatment:
Non-treated
Tissue Culture
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Surface Treatment
01670254 Receiver Plate Tissue Culture
01670252 Receiver Plate Non-treated
01670253 Insert with Permeable Membrane Tissue Culture
01670255 Insert with Transparent Membrane Tissue Culture
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01670254 Supplier Greiner Bio-One Supplier No. 655167
Only null left

Thincert™ 96 Well HTS inserts are indispensable tools for high-throughput screening and assays. Designed for precision, they enable researchers to elevate the quality and efficiency of their work.

  • Optimal tool for generating tissue models (e.g., endothelia and epithelia) for transport, uptake, or co-culture studies in submersed as well as air-lift culture with an automation-friendly format
  • By combining high membrane permeability and best possible transparency, Greiner Bio-One is offering a unique 96 well insert version, exclusively designed for applications that require both high diffusion rates as well as microscopic readouts.
  • Special pore arrangement and a high pore density of 2 x 107/cm2.
  • Patented assembly technology: robust and reliable bonding of membrane 
  • Excellent membrane flatness: optimal and reproducible cell culture conditions 
  • Optimized fit: prevention of media wicking
  • Enlarged access ports: convenient access for pipettes, robots and TEER electrodes

Specifications

Barcode No Barcode
Format Standard
Array 8 x 12
Color Clear
Product Type Receiver Plate
Material Polycarbonate
Volume (Metric) Well 1.25 to 3.12 μL
No. of Wells 96
Volume (Metric) Working 120 to 300 μL
Sterility Sterile
Surface Treatment Tissue Culture
Well Shape Flat Bottom (F-well)
Show More Show Less