missing translation for 'onlineSavingsMsg'
Learn More

Wilmad-LabGlass™ Thin Walled High Throughput NMR Tubes

Catalog No. 16800161
Change view
Click to view available options
Length (Metric):
103.5 mm
178 mm
203 mm
Field Strength:
600 MHz
High-throughput
Quantity:
100/Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Field Strength Quantity
16800161 103.5 mm 600 MHz 100/Pk.
16800495 178 mm High-throughput 100/Pk.
16800208 178 mm 600 MHz 100/Pk.
16800497 203 mm High-throughput 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 16800161 Supplier Wilmad-LabGlass™ Supplier No. 660000061
Get Up to $500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Possess an average camber of 60 microns to guarantee the clarity of the spectrum for small molecule (MW<250) samples up to 600 MHz. Wilmad Thin Walled High Throughput NMR Tubes are designed for routine use in most low to mid field NMR spectrometers.

  • Camber and concentricity values listed are the Total Indicator Reading (TIR)
  • Designed for routine use in most low to mid field NMR spectrometers
  • One of the best OD tolerances in the industry
  • Guaranteed to fit tightly in spinner turbine
  • Made from ASTM E438 Type 1 Class B glass
  • 100% inspected for surface defects and physical dimension to ensure the success of your experiments
  • WG-1000-4-SJ and WG-1000-7-SJ are supplied with authentic Bruker SampleJet caps, all others come with a disposable NMR cap
Note: For NMR experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, please refer to Wilmad Precision NMR Tubes.

Specifications

Length (Metric) 103.5 mm
Camber (Metric) 60 μm
Material ASTM E438 Type 1 Class B Glass
Product Type NMR Tube
Closure Type Bruker SampleJet Cap
Diameter (Metric) Outer 5 mm
Field Strength 600 MHz
Length (English) 4 in.
Thickness (Metric) Wall 0.43 mm
Quantity 100/Pk.
Model WG-1000-4-SJ
Show More Show Less