missing translation for 'onlineSavingsMsg'
Learn More

Thieno[3,2-b]thiophene-2-carbonitrile 98.0+%, TCI America™

Catalog No. T3221200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3221200MG 200 mg
T32211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3221200MG Supplier TCI America Supplier No. T3221200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40985-58-8
Molecular Formula C7H3NS2
Molecular Weight (g/mol) 165.228
MDL Number MFCD08059461
InChI Key UPRBPGCXCNBTQH-UHFFFAOYSA-N
Synonym 2-Cyanothieno[3,2-b]thiophene
PubChem CID 55252708
IUPAC Name thieno[3,2-b]thiophene-5-carbonitrile
SMILES C1=CSC2=C1SC(=C2)C#N

Specifications

Melting Point 50°C
Color White-Yellow
Quantity 200 mg
Formula Weight 165.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Thieno[3,2-b]thiophene-2-carbonitrile