missing translation for 'onlineSavingsMsg'
Learn More

Thieno[2,3-b]thiophene 97.0+%, TCI America™

Catalog No. T27291G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27291G 1 g
T27295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27291G Supplier TCI America Supplier No. T27291G

Chemical Identifiers

CAS 250-84-0
Molecular Formula C6H4S2
Molecular Weight (g/mol) 140.218
MDL Number MFCD02824126
InChI Key YHBTXTFFTYXOFV-UHFFFAOYSA-N
Synonym 1,6-Dithiapentalene
PubChem CID 136062
IUPAC Name thieno[2,3-b]thiophene
SMILES C1=CSC2=C1C=CS2

Specifications

Melting Point 7°C
Color Yellow
Boiling Point 226°C
Quantity 1 g
Formula Weight 140.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Thieno[2,3-b]thiophene