missing translation for 'onlineSavingsMsg'
Learn More

Thieno[2,3-b]pyridine-2-carboxylic acid, 95%, Thermo Scientific™

Catalog No. p-3176173
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC05601CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC05601CB Supplier Thermo Scientific Chemicals Supplier No. CC05601CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 59944-76-2
Molecular Formula C8H5NO2S
Molecular Weight (g/mol) 179.193
MDL Number MFCD07371381
InChI Key XGCSHAYMNOFFNA-UHFFFAOYSA-N
Synonym thieno 2,3-b pyridine-2-carboxylic acid, thieno 2,3-b pyridine-2-carboxylicacid, 2-carboxythieno 2,3-b pyridine, thiopheno 2,3-b pyridine-2-carboxylic acid, hieno 2,3-b pyridine-2-carboxylic acid
PubChem CID 7162095
IUPAC Name thieno[2,3-b]pyridine-2-carboxylic acid
SMILES C1=CC2=C(N=C1)SC(=C2)C(=O)O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 179.2
Percent Purity 95%
Chemical Name or Material Thieno[2, 3-b]pyridine-2-carboxylic acid
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.