missing translation for 'onlineSavingsMsg'
Learn More

Thieno[2,3-b]pyrazine-6-carboxylic acid, 90%, Thermo Scientific™

Catalog No. CC31501CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC31501CB 250mg Amber Glass Bottle
CC31501DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC31501CB Supplier Thermo Scientific Chemicals Supplier No. CC31501CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 59944-79-5
Molecular Formula C7H4N2O2S
Molecular Weight (g/mol) 180.181
InChI Key MEHCDDSVVYRWJT-UHFFFAOYSA-N
Synonym thieno 2,3-b pyrazine-6-carboxylic acid, pubchem9882, thieno 2,3-b pyrazine-6-carboxylicacid, 6-thieno 2,3-b pyrazinecarboxylic acid, thieno 3,2-b pyrazine-6-carboxylic acid, thiopheno 2,3-b pyrazine-6-carboxylic acid
PubChem CID 7164578
IUPAC Name thieno[2,3-b]pyrazine-6-carboxylic acid
SMILES C1=CN=C2C(=N1)C=C(S2)C(=O)O

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 180.19
Percent Purity 90%
Chemical Name or Material Thieno[2, 3-b]pyrazine-6-carboxylic acid