missing translation for 'onlineSavingsMsg'
Learn More

Thiazole-4-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7137245
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24941G 1 g
T24945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24941G Supplier TCI America Supplier No. T24941G

Chemical Identifiers

CAS 3973-08-8
Molecular Formula C4H3NO2S
Molecular Weight (g/mol) 129.13
MDL Number MFCD00623589
InChI Key HMVYYTRDXNKRBQ-UHFFFAOYSA-N
Synonym thiazole-4-carboxylic acid, 4-thiazolecarboxylic acid, 4-thiazolecarboxylicacid, 4-carboxythiazole, 4-carboxy-1,3-thiazole, thiazole 4-carboxylic acid, acmc-2097eb, thiazol-4-carboxylic, ksc225i1n
PubChem CID 304271
ChEBI CHEBI:46230
IUPAC Name 1,3-thiazole-4-carboxylic acid
SMILES C1=C(N=CS1)C(=O)O

Specifications

Melting Point 197°C
Color White-Yellow
Quantity 1 g
Formula Weight 129.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Thiazole-4-carboxylic Acid