missing translation for 'onlineSavingsMsg'
Learn More

Thiamine Thiocarbamate, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023012
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023012 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023012 Supplier Merck Emd Millipore Supplier No. PHR2207100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

  • Grade: Certified Reference Material
  • Linear Formula: C14H20N4O3S2
  • Formula Weight: 356.46
  • CAS Registry Number: 89285-03-0
  • Synonyms: (3 RS)-3-[[[(4-Amino-2-methylpyrimidin-5-yl)methyl]thiocarbamoyl]sulfanyl]-4-oxopentyl acetate (ketodithiocarbamate); Thiamine Impurity H
  • Pharmaceutical secondary standard for quality control applications.

Specifications

Chemical Name or Material Thiamine Thiocarbamate
CAS 89285-03-0
Physical Form Solid
Quantity 100 mg
Molecular Formula C14H20N4O3S2
Linear Formula C14H20N4O3S2
UN Number NONH for all modes of transport
Synonym (3 RS)-3-[[[(4-Amino-2-methylpyrimidin-5-yl)methyl]thiocarbamoyl]sulfanyl]-4-oxopentyl acetate (ketodithiocarbamate); Thiamine Impurity H
InChI Formula InChI=1S/C14H20N4O3S2/c1-9(19)5-6-23-14(22)16-7-10-8-17-12(15)11(2)18-13(10)21-4/h8,18H,5-7H2,1-4H3,(H2,15,17)(H,16,22)
InChI Key WJNDWTYYIDYJRS-UHFFFAOYSA-N
SMILES CC1=NC=C(C(=N1)CN)C(=O)SCSCC(CC(=O)C)OC(=O)C
IUPAC Name 3-[[[(4-amino-2-methylpyrimidin-5-yl)methyl]carbamothioyl]sulfanyl]-4-oxopentyl acetate
Molecular Weight (g/mol) 356.46
Formula Weight 356.46
Grade Certified Reference Material
Product Line Supelco
Show More Show Less