missing translation for 'onlineSavingsMsg'
Learn More

Thiamine Hydrochloride, Powder, MP Biomedicals

Catalog No. p-3179228
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10302880 100 g
ICN10302883 250 g
ICN10302891 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10302880 Supplier MP Biomedicals Inc Supplier No. 0210302880
Only null left

Thiamine (Vitamin B1) is one of the essential vitamins and is required for carbohydrate metabolism.

Thiamine hydrochloride is used as a food-additive to add a brothy/meaty flavor to gravies or soups. It is also used as food supplement. Used in fluorometric determination of mercury.

Chemical Identifiers

CAS 67-03-8
Molecular Formula C12H17ClN4OS·HCl
Molecular Weight (g/mol) 337.263
MDL Number MFCD00012780
InChI Key UIERGBJEBXXIGO-UHFFFAOYSA-N
Synonym Aneurine hydrochloride, Vitamin B1 hydrochloride, Vitamin B1 HCl
PubChem CID 10762
SMILES [O-][N+]([O-])=O.CC1=C(CCO)[S+]=CN1CC1=CN=C(C)N=C1N

Specifications

Quantity 100 g
Beilstein 3851771
IUPAC Name 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-3H-1λ4,3-thiazol-1-ylium nitrate
Formula Weight 337.3
Physical Form Powder
Chemical Name or Material Thiamine Hydrochloride, Food Supplement
EINECSNumber 200-641-8
storageNote1 Store Desiccated
Recommended Storage Store at room temperature.