missing translation for 'onlineSavingsMsg'
Learn More

Thiamine Hydrochloride, Powder, MP Biomedicals

Catalog No. ICN10302880
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10302880 100 g
ICN10302883 250 g
ICN10302891 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10302880 Supplier MP Biomedicals Inc Supplier No. 0210302880

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thiamine (Vitamin B1) is one of the essential vitamins and is required for carbohydrate metabolism.

Thiamine hydrochloride is used as a food-additive to add a brothy/meaty flavor to gravies or soups. It is also used as food supplement. Used in fluorometric determination of mercury.

Chemical Identifiers

CAS 67-03-8
Molecular Formula C12H17ClN4OS·HCl
Molecular Weight (g/mol) 337.263
MDL Number MFCD00012780
InChI Key UIERGBJEBXXIGO-UHFFFAOYSA-N
Synonym Aneurine hydrochloride, Vitamin B1 hydrochloride, Vitamin B1 HCl
PubChem CID 10762
SMILES [O-][N+]([O-])=O.CC1=C(CCO)[S+]=CN1CC1=CN=C(C)N=C1N

Specifications

Quantity 100 g
Beilstein 3851771
IUPAC Name 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-3H-1λ4,3-thiazol-1-ylium nitrate
Formula Weight 337.3
Physical Form Powder
Chemical Name or Material Thiamine Hydrochloride, Food Supplement
EINECSNumber 200-641-8
storageNote1 Store Desiccated
Recommended Storage Store at room temperature.