missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thiamine hydrochloride, 98.5-101.5%

Catalog No. AC148990010
Change view
Click to view available options
Quantity:
10 g
100 g
1 kg
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC148990010 1 kg Glass Bottle
AC148990100 10 g Glass Bottle
AC148991000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC148990010 Supplier Thermo Scientific Chemicals Supplier No. 148990010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thiamine hydrochloride, 98.5-101.5%, C12H17ClN4OS.HCl, CAS Number-67-03-8, aneurine hydrochloride, berin, thiamin chloride, thiamine chloride hydrochloride, thiamine dichloride, thiamine hcl, thiamine hydrochloride, thiaminium chloride, trophite, vitamin b1 hydrochloride, 1kg, 98.5 to 101.5% | CAS: 67-03-8 | C12H17ClN4OS·HCl | 337.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67-03-8
Molecular Formula C12H17ClN4OS·HCl
Molecular Weight (g/mol) 337.26
MDL Number MFCD00012780
InChI Key DPJRMOMPQZCRJU-UHFFFAOYSA-M
Synonym thiamine hydrochloride, vitamin b1 hydrochloride, thiamine hcl, aneurine hydrochloride, berin, thiamine chloride hydrochloride, trophite, thiamin chloride, thiamine dichloride, thiaminium chloride
PubChem CID 6202
ChEBI CHEBI:49105
IUPAC Name 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride
SMILES CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]

Specifications

Melting Point 250.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5 to 101.5%
Packaging Glass Bottle
Quantity 1 kg
Beilstein 27, IV, 1766
Merck Index 15, 9447
Solubility Information Solubility in water: 1 g/mL. Other solubilities: 1 g/18 mL glycerol - 1 g/100 mL 95% alcohol, 1 g/315 mL abs. alcohol, soluble in propylene glycol, practically insoluble in hexane, chloroform, insoluble in benzene, diethyl ether
Formula Weight 337.26
Percent Purity 98.5-101.5%
Physical Form Crystalline Powder
Chemical Name or Material Thiamine hydrochloride, 98.5-101.5%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-641-8
RTECSNumber XI7350000
TSCA TSCA

RUO – Research Use Only