missing translation for 'onlineSavingsMsg'
Learn More

Thiabendazole, 98+%

Catalog No. AAJ6000909
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6000909 10 g
AAJ6000922 100 g
AAJ6000936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6000909 Supplier Thermo Scientific Chemicals Supplier No. J6000909
Only null left

Fungicide and antihelminthic | CAS: 148-79-8 | C10H7N3S | 201.247 g/mol

Thiabendazole, a metal chelator and helminth-specific fumarate reductase inhibitor, is used primarily as an anthelmintic to control infections by helminth organisms. It is also used to control mold and fungal growth in plants. Also used to treat parasitic infections in humans. It is used clinically to treat threadworm, cutaneous larva migrans, visceral larva migrans, and trichinosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in methanol and dimethyl sulfoxide.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 148-79-8
Molecular Formula C10H7N3S
Molecular Weight (g/mol) 201.247
MDL Number MFCD00005587
InChI Key WJCNZQLZVWNLKY-UHFFFAOYSA-N
Synonym thiabendazole, tiabendazole, mintezol, equizole, tiabendazol, mintesol, omnizole, thibenzole, thiabendazol, bovizole
PubChem CID 5430
ChEBI CHEBI:45979
IUPAC Name 4-(1H-benzimidazol-2-yl)-1,3-thiazole
SMILES C1=CC=C2C(=C1)NC(=N2)C3=CSC=N3

Specifications

Melting Point 304°C to 305°C
Color White
Quantity 10 g
UN Number UN3077
Beilstein 611403
Merck Index 14,9289
Solubility Information Soluble in methanol and dimethyl sulfoxide.
Formula Weight 201.25
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Thiabendazole
Show More Show Less
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 205-725-8
RTECSNumber DE0700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only