Learn More
Laboratory Craftsmen Inc Thermowell C3AM 50mL Heating Mantle, 100 W, CSA Certified, Low Profile Design
Catalog No. NC0023647
Encompass
The THERMOWELL C3AM 50mL Heating Mantle by Laboratory Craftsmen Inc. provides reliable uniform heating for laboratory applications, compatible with 50 mL round-bottom flasks. It features a robust design with a ceramic heat cavity and embedded nichrome wire, allowing for various heating methods like air or sand baths. Its low-profile design enhances magnetic stirring compatibility, making it a versatile choice for distillations and extractions.
Catalog No. NC0023647 Supplier Laboratory Craftsmen Inc Supplier No. C3AM 50ML
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The THERMOWELL C3AM 50mL Heating Mantle by Laboratory Craftsmen Inc. provides reliable uniform heating for laboratory applications, compatible with 50 mL round-bottom flasks. It features a robust design with a ceramic heat cavity and embedded nichrome wire, allowing for various heating methods like air or sand baths. Its low-profile design enhances magnetic stirring compatibility, making it a versatile choice for distillations and extractions.