missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Thermowell™ Gold 96-Well Polypropylene PCR Microplates

Catalog No. 07201217
Change view
Click to view available options
Color:
Clear
Skirt Style:
Elevated
Full
Half
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Skirt Style
07201217 Clear Elevated
07201216 Clear Half
07201215 Clear Full
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07201217 Supplier Corning™ Supplier No. 3754

Available in full, half, and elevated skirt styles

Corning Thermowell GOLD 96-well PCR microplates are offered in four formats to ensure maximum flexibility and a perfect match for your applications. The original Thermowell 96-well PCR microplates are universal fit and can be cut into 3 x 8 well segments.

Specifications

Certifications/Compliance DNase, RNase-free
Format Standard
Lid Without Lid
Array 8 x 12
Packaging 5 inner bags of 10
Color Clear
Skirt Style Elevated
Material Polypropylene
Volume (Metric) Well 200 μL
No. of Wells 96
Sterility Non-sterile
Well Shape Round
For Use With (Application) For automation, RT-PCR applications
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.
Well Color Clear
Show More Show Less