missing translation for 'onlineSavingsMsg'
Learn More

Sonoco™ ThermoSafe U-tek™ Ice

Catalog No. 03600107
Change view
Click to view available options
Dimensions (L x W):
16 x 14.7 in. (40.6 x 37.3 cm)
3 x 1.5 in. (7.6 x 3.8 cm)
Type:
U-tek Single
U-tek Square Mat
Quantity:
1 Cs.
28 Sheets
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Type Quantity
03600107 16 x 14.7 in. (40.6 x 37.3 cm) U-tek Square Mat 28 Sheets
035283 3 x 1.5 in. (7.6 x 3.8 cm) U-tek Single 1 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03600107 Supplier Sonoco™ Supplier No. 405SQ

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cushions fragile shipments, and won’t dilute ice baths or damage samples. Remains frozen longer than regular ice, without the melting mess.

  • Can be purchased as a 34ft roll,or as a 16 X 14 inch mat, or individual packets
  • May be refrozen and reused, withstands pressures in excess of 60 lb per square inch for five minutes or longer 
  • Refrigerant contains neutral pH distilled and deionized water, and proprietary materials certified by the U.S. Food and Drug Administration as food grade chemicals
  • Freezing Point: -1 deg. C/+30 deg

Specifications

Quantity 28 Sheets
Type U-tek Square Mat
Dimensions (L x W) 16 x 14.7 in. (40.6 x 37.3 cm)
Material Phase Change Material
Freezing Point -1° C
For Use With (Application) Ideal for products requiring frozen temperatures or extreme cold that may not tolerate dry ice or Carbon dioxide