Learn More
Sonoco™ ThermoSafe PolarPack™ Standard
PolarPack refrigerants provide thermal solutions for temperature critical packages during transit. Available in a broad selection of styles and a multitude of sizes, our refrigerants are designed to provide reliable temperature protection for a vast array of temperature-sensitive products and packages.
Supplier:  Sonoco™ PP3
 View more versions of this product
Catalog No. 03532100
Only null left
Description
Description
  • Thermal solutions for temperature critical packages during transit
  • Gel will phase change at 0° C.
  • condition coolant in freezer before shipments.
  • refrigeration for transport of food, pharmaceuticals and medical products. 
  • For frozen or refrigerated shipments.
  • Alternative to wet ice.
  • Available in several sizes
  • Cold shipping supplies .
Specifications
Chemical Identifiers
0°C
Specifications
3 oz.
Polyethylene
For shipping temperature critical packages during transit
3.75 x 4 x 0.62 in.
0°C
Product Suggestions
Product Suggestions