Learn More
Sonoco™ ThermoSafe Nomadic™ Pre-Qualified Shippers: Accessories
Catalog No. 22031871
Change view
Click to view available options
Type:
Shipper; 2-8 °C; 48-hour
Catalog No. Type
22031871 Shipper; 2-8 °C; 48-hour
1 options
Catalog No. 22031871 Supplier Sonoco™ Supplier No. E280C
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
24 oz. Polar pack tough pack (8), 20 oz. Polar pack standard gel pack (3), corrugated outer corepack (1 or 2), double-wall laminated pads (1), corrugated inner corepack (1)
For protection of refrigerated and frozen pharmaceuticals and biological samples and specimens
  • Meet toughest ambient profiles of any premium shipper in the industry
  • Multicomponent packing system to ensure exceptional temperature protection and durability
  • Model E48UPS:
    • Complies with international clean air regulations, CFC- and HCFC-free
    • Qualified for maintaining payloads of a single 1mL vial to 3L of liquid
    • Reusable
    • Replaceable components available separately
    • With two double-wall laminated pads
  • Model E24AUPS:
    • Specifically engineered for quick trips of 24 hours or less
    • Lightweight and cost-effective EPS inner shell reduces overnight shipping costs
    • Superior insulation power of 2 in. thick walls
    • With one double-wall laminated pad
Order Info
Container, corepacks and gels shipped in a single overpack.
Specifications
Applications For refrigerated, frozen pharmaceutical and biological specimens
Type Shipper; 2-8 °C; 48-hour