missing translation for 'onlineSavingsMsg'
Learn More

Sonoco™ ThermoSafe Metalized Bubble Pouches

Catalog No. 03600159
Change view
Click to view available options
Length (English) Exterior:
8 in.
12 in.
18 in.
Width (English) Exterior:
11 in.
17 in.
23 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Exterior Width (English) Exterior
03600159 18 in. 23 in.
03600154 8 in. 11 in.
03600158 12 in. 23 in.
03600156 12 in. 17 in.
03600155 12 in. 11 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03600159 Supplier Sonoco™ Supplier No. MP1823

Lightweight, economical EPS shippers provide maximum breakage protection for known non-hazardous materials.

  • Accommodates VacuTainer™, Microtainer™, and other collection/transport tubes
  • Available in multi-tube, slide, and tube/slide combinations
  • Lightweight polystyrene foam construction saves postage
  • Reusable and recyclable

Specifications

Quantity 100 Pouches/Pk.
Length (English) Exterior 18 in.
Width (English) Exterior 23 in.
Includes 2 in. Self sealing lip and tape
Type Bubble Pouch
Dimensions (L x W) 18 x 23 in.
Material 0.187 in. thick metallized bubble