missing translation for 'onlineSavingsMsg'
Learn More

Sonoco™ ThermoSafe LabMailer Specimen Mailer

Catalog No. p-150274
Change view
Click to view available options
Type:
Mailer 335 for Specimens
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
035295A Mailer 335 for Specimens
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 035295A Supplier Sonoco™ Supplier No. 335
Only null left

Lightweight, economical lab mailer shippers

  • Hold screw-capped or rubber-capped tubes or vials and swabs
  • Provide maximum breakage protection for known nonhazardous materials
  • Protect frozen, refrigerated, and ambient specimens required for compliance, analysis, testing, research and development, public health, industrial hygiene and environmental use

Additional packaging may be required to conform to CFR72.2 specification (See Packaging Accessories Kit Mfr. No. 471).

Specifications

Type Mailer 335 for Specimens
Dimensions (L x W x H) Exterior 7 x 2.5 x 2.5 in.
Dimensions (D x W x H) Interior 5.875 x 1.5 x 1.5 in.
Description Holds specimen tubes, vials, swabs. Must order Model 336 for mailing sleeve.
Dimensions (LxWxH) Outside: 7 x 2.5 x 2.5 in.; Cavity: 5.875 x 1.5 x 1.5 in.