Learn More
Sonoco™ ThermoSafe LabMailer Sleeves for Tube and Slide Mailers
Catalog No. 0352535
Change view
Click to view available options
For Use With (Equipment):
Mailer 339, 364 and 394
Mailer 407
Material:
18-pt White Paperboard
Sturdy White Cardboard
Includes:
Blood Legend Label
Plain Label
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Material Includes
0352535 Mailer 407 18-pt White Paperboard Plain Label
03528A Mailer 339, 364 and 394 Sturdy White Cardboard Plain Label
0353125 Mailer 339, 364 and 394 Sturdy White Cardboard Blood Legend Label
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 0352535 Supplier Sonoco™ Supplier No. TS408
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Sturdy white cardboard with reverse-tuck closures
03-531-25 and 03-530-10 have printed blood legend; all others are plain
Specifications
Includes Plain Label
For Use With (Equipment) Mailer 407
Type Tube and Slide Mailer Sleeve
Dimensions (LxWxH) 5.63 x 1.94 x 6.5 in.
Material 18-pt White Paperboard