missing translation for 'onlineSavingsMsg'
Learn More

Sonoco™ ThermoSafe LabMailer Shipping Containers

Catalog No. 03531
Change view
Click to view available options
Type:
Mailer Model 312
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
03531 Mailer Model 312
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03531 Supplier Sonoco™ Supplier No. TS312

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Accommodates a variety of tubes, vials, cans and bottles

  • 03-525-23B: Mailing sleeve for Model 03-525-34B
  • 03-525-34B: Sturdy 0.5 in. (1.3 cm) walls protect against shock and breakage
  • Foam mailer and paperboard mailing sleeve sold separately
  • Interior dimensions: 4L × 3.25W × 3.25 in.H (10.2 × 8.3 × 8.3cm)

Specifications

Quantity 12/Cs.
Applications Maximum breakage protection for shipment of known non-hazardous materials
Height (English) Exterior 7.12 in.
Type Mailer Model 312
Dimensions (L x W x H) Exterior 10 x 10 x 7.5 in.
Height (Metric) Exterior 18 cm
Description Holds Tubes, Vials, Ampules
Dimensions (LxWxH) 9.87 x 8.87 x 7.12 in. (25 x 25 x 18 cm)
Material Reusable/recyclable polystyrene foam