Learn More
Sonoco™ ThermoSafe Diagnostic Shipper, Three Tube Mailer System
Catalog No. p-150324
Change view
Click to view available options
Type:
Complete Three-Tube Mailer System
Foam Mailer Only (no tubes)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
03527D Complete Three-Tube Mailer System
0352826 Foam Mailer Only (no tubes)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03527D Supplier Sonoco™ Supplier No. TS473
Only null left
Mailer secures up to three 10mL tubes
  • 95kPa certified receptacles (tubes or bags depending on kit)
  • Ready-to-use
  • Sufficient absorbent material to absorb maximum capacity of liquid
  • IATA Packing Instruction 650, Biological Substance, Category B label
Compliance
Meets U.S. Postal Regulations, DOT regulations, and IATA Packaging Instruction 650; tubes and shipping system validated for the transport of diagnostic samples.
Specifications
Includes Three 10mL vial tubes, EPS foam mailer with lid, absorbent material, water-resistant tape, pressure-lock plastic bag, 200 lb. test corrugated shipping carton with lock tab and instructions
Certifications/Compliance Meets U.S. Postal Regulations, DOT regulations, and IATA Packaging Instruction 650; tubes and shipping system validated for the transport of diagnostic samples
Type Complete Three-Tube Mailer System
Material Expanded Polystyrene Foam