Learn More
Sonoco™ ThermoSafe Diagnostic Shipper, Eight Tube Mailing System, Complete Mailer System
Catalog No. 03527C
Change view
Click to view available options
Type:
Diagnostic Shipper
Catalog No. Type
03527C Diagnostic Shipper
1 options
Catalog No. 03527C Supplier Sonoco™ Supplier No. TS472
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
Complete set (Cat. No. 03-527C): 10mL vial tubes, foam mailer with lid, absorbent material, water-resistant tape, pressure-lock plastic bag, 200 lb. test corrugated shipping carton with antitamper locking tabs and instructions. Cat. No. 03-527B includes eight-tube safety mailer only.
Leak-resistant, EPS foam mailer holds 30mL bottle, eight 10mL tubes, materials for insurance testing and other paperwork
  • Tight-fitting plug cover accepts required absorbent material
  • Validated for the transport of diagnostic samples
Compliance
Meets U.S. Postal Regulations, DOT regulations, and IATA Packaging Instruction 650; tubes and shipping system validated for the transport of diagnostic samples
Specifications
Includes 10mL vial tubes, foam mailer with lid, absorbent material, water-resistant tape, pressure-lock plastic bag, 200 lb. test corrugated shipping carton with antitamper locking tabs and instructions
Certifications/Compliance Meets U.S. Postal Regulations, DOT regulations, and IATA Packaging Instruction 650; tubes and shipping system validated for the transport of diagnostic samples
Applications Diagnostic materials
Type Diagnostic Shipper
Capacity 30 mL
Material Expanded Polystyrene Foam and Cardboard, Plastic Tubes