missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ Thermolite Plus Septa
Ultra-low bleed minimizes background signal
Specifications
| For Use With (Application) | Chromatography |
|---|---|
| Temperature (Metric) Max | 350°C |
| Type | Thermolite Plus Septa |
Description
- Usable to 350°C inlet temperature (except 17 mm inlets max temp 300°C)
- New plasma coating eliminates sticking in injection port
- Precision molding ensures consistent, accurate fit
- Septa have a CenterGuide design to minimize coring
- (Not available on Shimadzu plug, 6, 7, 8, 9.5, and 10 mm)
- 5 mm septa partially predrilled for improved puncturability
- Preconditioned and ready to use
- Packaged in ultra-clean blister packs
- Each batch GC-FID tested
Note:
- Due to differences in inlet design, actual septum temp for a given inlet set point can vary by manufacturer
- Recommended using only BTO septa in older Thermo TRACE and Focus GCs
- Does not apply for Thermo TRACE 1300/1310 GCs
- Septa can be used only to max recommended temp
Specifications
| Chromatography | |
| 350°C | |
| Thermolite Plus Septa |