missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Thermo Sequenase DNA Polymerase

Catalog No. 09928126
Change view
Click to view available options
Quantity:
1,000 Units
10,000 Units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09928126 1,000 Units
09928127 10,000 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09928126 Supplier Cytiva Supplier No. E79000Y
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Novel thermostable DNA polymerase that uses dideoxynucleotide triphosphates (ddNTPs) as readily as deoxynucleotide (dNTP) substrates

  • Uniform and easy-to-read band patterns
  • Better incorporation of ddNTPs helps avoid secondary structures that can cause unreadable sequences
  • Read-through of difficult sequences (e.g., GC-rich), due to the ability of thermostable enzymes to function at elevated temperature used for sequencing
  • ddNTPs are chain-elongating inhibitors of DNA polymerase, which are used in the Sanger method for DNA sequencing
  • Each nucleotide base of this particular type has a probability of being bonded not to a deoxynucleotide but rather a dideoxynucleotide, which ends chain elongation
  • An excellent choice for cycle sequencing where template quantity limitations preclude the use of T7 Sequenase DNA polymerase
  • Can also be used in many SNP genotyping, genotyping by sequencing, or genome sequencing applications where primer extension is employed

Specifications

Content And Storage -20°C, Solution in 20 mM Tris-HCl, pH 8.5, 0.1 mM EDTA, 0.5% Tween 20 (v/v), 0.5% Nonidet P-40 (v/v), 1 mM DTT, 100 mM KCl, 50% glycerol
Description Thermo Sequenase DNA Polymerase
Quantity 1,000 Units
For Use With (Application) Sequencing, Genotyping
No. of Reactions 32 units/μL
Includes Thermo Sequenase with Thermoplasma Acidophilum Inorganic Pyrophosphatase (TAP), Dilution Buffer, and Reaction Buffer
Source E. coli clones that overproduce the protein