missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Workstation/Stacking System for Tubes/Plates

Catalog No. AB0538
Change view
Click to view available options
Quantity:
10 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB0538 10 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB0538 Supplier Thermo Scientific™ Supplier No. AB0538

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Convenient platform for tube handling and reagent dispensing .

Highlights

  • Convenient platform for tube handling and reagent dispensing
  • Holds:
    - 96 x 0.2ml V-bottom tubes
    - 96 x 0.3ml U-bottom Omnistrips
    - Thermo-Tube Plates
    - Thermo-Fast 96 Non-Skirted Plate
    - Thermo-Fast 96 Semi-Skirted Plate
    - 48 x 0.5ml Thermo-Tubes
  • Multiple workstations can be securely stacked
  • Can be stored at -190°C

Hazardous: No
Storage Condition: Ambient

Specifications

For Use With (Equipment) Real-Time PCR Systems or Thermal Cyclers
Quantity 10 units