Learn More
Thermo Scientific™ VersiCap Mat, 96-well, flat cap strips
Description
Thermo Scientific VersiCap Mats are unique, with each strip of eight caps being linked to each other forming the familiar 12 x 8 or 96-well ANSI format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.
The Thermo Scientific VersiCap Mats are unique, with each strip of eight caps being linked to each other forming the familiar 12 x 8 or 96-well ANSI format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.
Highlights
- Multiple cap strips can be applied at the same time when sealing qPCR plates, thereby reducing setup time
- Remove the need to cut up adhesive seals
- Suitable for use with all Thermo Scientific 96-well PCR plates as well as standard and low profile strip tubes
- PCR
- qPCR/Real-Time PCR
Autoclavable Resistance: Successfully Tested
DMSO (100%) Resistance: Successfully Tested
Ethanol Resistance: Successfully Tested
Gamma Irradiation Resistance: Successfully Tested
Hazardous: No
Isopropanol Resistance: Successfully Tested
Long Term Storage: Successfully Tested
PCR (including waterbath): Successfully Tested
Peelable: Successfully Tested
Pierceable: Not Recommended
qPCR:Ultra Clear caps: Successfully Tested
Domed or Flat caps: Not Recommended
Resealable: Successfully Tested
Sealing Temperature Range: −20°C to +120°C
Shelf Life: 60 Months
Shipping Condition: Ambient
Storage Condition: Ambient
UV Irradiation Resistance: Successfully Tested
- PCR
- qPCR/Real-Time PCR
Certifications
Certified free from human DNA, RNases, and DNases
Specifications
Specifications
| Color | Clear |
| Material | Polypropylene |
| No. of Wells | 96 |
| No. per Case | 25 |
| Quantity | 25 mats |
| Format | Strips of 8 Flat Caps |
| Product Type | PCR Plate Cap Strip |
| Product Line | VersiPlate |
| Certified RNase/DNase-free | Yes |
| Temperature Range (Metric) | -20°C to +120°C |
| Show More |