missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TX-400 Rotor Adapters

Catalog No. 75003825
Change view
Click to view available options
For Use With (Equipment):
TX-400 Swinging Bucket Rotor
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
75003825 TX-400 Swinging Bucket Rotor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 75003825 Supplier Thermo Scientific™ Supplier No. 75003825

For use with TX-400 Swinging Bucket Rotor

The adapters enable rotor to accept bottles up to 250mL and tubes to 100mL.

  • Maximum adapter capacity: 4 x 250mL (4 x 400mL with 400mL polypropylene bio-bottles)
  • Maximum adapter O.D. x L: 62 x 125mm (80 x 125mm with 400mL bio-bottles)

Specifications

Type Rotor Adapter
Capacity 4.5/6 mL
Max. Tube Diameter 13 mm
Max. Tube Length 110 mm
For Use With (Equipment) TX-400 Swinging Bucket Rotor
Tube Size 4.5/6 mL