Learn More
Thermo Scientific™ TX-100/100S Clinical Swinging Bucket Rotor Adapters
Catalog No. 11172288
Change view
Click to view available options
Capacity:
1.5/2 mL Microtube
3 mL Blood Tube
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
11172288 1.5/2 mL Microtube
05376246 3 mL Blood Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11172288 Supplier Thermo Scientific™ Supplier No. 11172288
Adapters for TX-100S Clinical Rotor and TX-100 Clinical Rotor
Specifications
Type Microtube Adapter
Tube Type Microtube
Capacity 1.5/2 mL Microtube
For Use With (Equipment) TX-100 and TX-100S clinical swing bucket rotor
Tube Size 1.5/2 mL