missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSX Series High-Performance Lab Refrigerators
GREENER_CHOICE

Catalog No. TSX5005SD
Change view
Click to view available options
Capacity (English):
11.5 cu. ft.
23 cu. ft.
29.2 cu. ft.
45.8 cu. ft.
51.1 cu. ft.
Door Style:
Double Glass
Double Solid
Glass
Sliding Glass
Solid
Plug Type:
NEMA 5-15
NEMA 6-15
Electrical Requirements:
115V, 60Hz
208-230V, 50/60Hz
208-230V, 60Hz
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Door Style Plug Type Electrical Requirements
TSX5005SD 51.1 cu. ft. Double Solid NEMA 6-15 208-230V, 60Hz
TSX2305GA 23 cu. ft. Glass NEMA 5-15 115V, 60Hz
TSX4505GD 45.8 cu. ft. Sliding Glass NEMA 6-15 208-230V, 60Hz
TSX5005GA 51.1 cu. ft. Double Glass NEMA 5-15 115V, 60Hz
TSX5005GD 51.1 cu. ft. Double Glass NEMA 6-15 208-230V, 60Hz
TSX5005SA 51.1 cu. ft. Double Solid NEMA 5-15 115V, 60Hz
TSX1205GA 11.5 cu. ft. Glass NEMA 5-15 115V, 60Hz
TSX1205GD 11.5 cu. ft. Glass NEMA 6-15 208-230V, 60Hz
TSX1205SA 11.5 cu. ft. Solid NEMA 5-15 115V, 60Hz
TSX1205SD 11.5 cu. ft. Solid NEMA 6-15 208-230V, 60Hz
TSX3005GA 29.2 cu. ft. Glass NEMA 5-15 115V, 60Hz
TSX3005GD 29.2 cu. ft. Glass NEMA 6-15 208-230V, 60Hz
TSX3005SA 29.2 cu. ft. Solid NEMA 5-15 115V, 60Hz
TSX3005SD 29.2 cu. ft. Solid NEMA 6-15 208-230V, 60Hz
TSX2305GD 23 cu. ft. Glass NEMA 6-15 208-230V, 50/60Hz
TSX2305SD 23 cu. ft. Solid NEMA 6-15 208-230V, 50/60Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. TSX5005SD Supplier Thermo Scientific™ Supplier No. TSX5005SD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Innovative refrigeration solutions for healthcare and research needs.

Thermo Scientific™ TSX Series high-performance lab refrigerators are designed with features that support sample protection and sustainability objectives for the storage of media, reagents, chemotherapy, pharmaceutical and other medical and laboratory-grade storage requiring 3° to 7°C. The Series' V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.

Featuring over 450W of reserve refrigeration capacity, the TSX series has the power to protect your product from temperature swings as a result of frequent and prolonged door access, large product loads, and other activities that introduce hear to the cabinet.

The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.

The drive for sample integrity and security

  • Designed to safely store media, reagents, chemotherapy, pharmaceutical and other medical and laboratory-grade products requiring a temperature range of 3°C to 7°C
  • Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Standard door locks provide comfort and assurance

Enabling compliance

  • Tested to GMP Clean Room Class A / ISO 5 & 6 (ISO EN 14644-1) with easy to clean surfaces support USP <797> and <800> hazardous drug compounding and other clean room environments
  • Tested and certified to conform with the design and safety standards of UL 471, 61010, and CE Directive 2011/65/EU and 2015/863

Designed for an ergonomic and comfortable experience

  • Ergonomically designed door handle with a soft-touch grip
  • The controller is angled to help prevent neck strain
  • Convenient, easy to clean and adjustable epoxy coated full and half shelves
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Bright, LED interior lighting
  • Self-closing multipaned glass or solid doors with 90° stop to assist with inventory loads

Verified sustainability

  • US EPA ENERGY STAR Certified with up to 40% less energy usage than conventional refrigerant models*
  • ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life. **
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
  • Manufactured in an award-winning, zero-waste to landfill facility

Ready for connectivity and monitoring

  • Thermo Scientific™ DeviceLink™ Connect ready, enabling you to have a connected experience with your unit so you always—and anywhere— know the safety of your samples and the performance of your refrigerator or freezer.
  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems

Make it your own

  • Alternate Door swing configuration and door types
  • Extra and Alternate wire baskets, shelves, and drawers
  • Free-standing or factory-installed chart recorders
  • Stainless Steel interior finish
  • E-Lock Adapter kits

Specifications

Type High-Performance Refrigerator
Capacity (English) 51.1 cu. ft.
Capacity (Metric) 1447 L
Temperature (Setpoint) 3°-7°C (5°C)
Refrigerant R290
Door Style Double Solid
No. of Doors 2
Shelves 8
Certifications/Compliance UL, cUL, ENERGYSTAR
Display Microprocessor
Product Line TSX Series
Warranty Warranty varies by country, please contact us for details
Defrost Automatic
Monitoring Options Alarm, Recorder
Chart Recorder Optional
Smart View Compatibility Smart-Vue Compatible
Dimensions (D x W x H) Interior 28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm)
Dimensions (L x W x H) Exterior 37.9 x 56.5 x 78.5 in. (96.2 x 143.5 x 199.4 cm)
Depth (English) Exterior 37.8 in.
Depth (English) Interior 28.5 in.
Depth (Metric) Exterior 96.0 cm
Depth (Metric) Interior 72.4 cm
Height (English) Exterior 78.5 in.
Height (English) Interior 58 in.
Height (Metric) Exterior 199.4 cm
Height (Metric) Interior 147.3 cm
Width (English) Exterior 56.5 in.
Width (English) Interior 52.5 in.
Width (Metric) Exterior 143.5 cm
Width (Metric) Interior 133.4 cm
Amperage 15 A (CB)
Plug Type NEMA 6-15
Voltage 208/230 V
Electrical Requirements 208-230V, 60Hz
Frequency 50/60 Hz
Green Features Energy efficient
Show More Show Less

*Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models.