Learn More
Thermo Scientific™ TSX Series High-Performance Lab Refrigerators

Innovative refrigeration solutions for healthcare and research needs.
Supplier: Thermo Scientific™ TSX4505GD
Description
Thermo Scientific™ TSX Series high-performance lab refrigerators are designed with features that support sample protection and sustainability objectives for the storage of media, reagents, chemotherapy, pharmaceutical and other medical and laboratory-grade storage requiring 3° to 7°C. The Series' V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
Featuring over 450W of reserve refrigeration capacity, the TSX series has the power to protect your product from temperature swings as a result of frequent and prolonged door access, large product loads, and other activities that introduce hear to the cabinet.
The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.
The drive for sample integrity and security
- Designed to safely store media, reagents, chemotherapy, pharmaceutical and other medical and laboratory-grade products requiring a temperature range of 3°C to 7°C
- Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
- Easy to use microprocessor control with interior light, setpoint security and alarms
- Standard door locks provide comfort and assurance
Enabling compliance
- Tested to GMP Clean Room Class A / ISO 5 & 6 (ISO EN 14644-1) with easy to clean surfaces support USP <797> and <800> hazardous drug compounding and other clean room environments
- Tested and certified to conform with the design and safety standards of UL 471, 61010, and CE Directive 2011/65/EU and 2015/863
Designed for an ergonomic and comfortable experience
- Ergonomically designed door handle with a soft-touch grip
- The controller is angled to help prevent neck strain
- Convenient, easy to clean and adjustable epoxy coated full and half shelves
- Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
- Bright, LED interior lighting
- Self-closing multipaned glass or solid doors with 90° stop to assist with inventory loads
Verified sustainability
- US EPA ENERGY STAR Certified with up to 40% less energy usage than conventional refrigerant models*
- ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life. **
- Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
- Manufactured in an award-winning, zero-waste to landfill facility
Ready for connectivity and monitoring
- Thermo Scientific™ DeviceLink™ Connect ready, enabling you to have a connected experience with your unit so you always—and anywhere— know the safety of your samples and the performance of your refrigerator or freezer.
- Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems
Make it your own
- Alternate Door swing configuration and door types
- Extra and Alternate wire baskets, shelves, and drawers
- Free-standing or factory-installed chart recorders
- Stainless Steel interior finish
- E-Lock Adapter kits
Chemical Identifiers
| R290 | |
| TSX Series | |
| Alarm, Recorder | |
| Smart-Vue Compatible |
| Microprocessor | |
| Automatic | |
| Optional |
Specifications
| High-Performance Refrigerator | |
| 1297 L | |
| R290 | |
| 2 | |
| UL, cUL, ENERGYSTAR | |
| TSX Series | |
| Automatic | |
| Optional | |
| 28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm) | |
| 36.6 in. | |
| 93.1 cm | |
| 78.5 in. | |
| 199.4 cm | |
| 56.5 in. | |
| 143.5 cm | |
| 15 A (CB) | |
| 208/230 V | |
| 50/60 Hz | |
| 611 lb. |
| 45.8 cu. ft. | |
| 3°-7°C (5°C) | |
| Sliding Glass | |
| 8 Wire | |
| Microprocessor | |
| Warranty varies by country, please contact us for details | |
| Alarm, Recorder | |
| Smart-Vue Compatible | |
| 36.6 x 56.5 x 78.5 in (93.1 x 143.5 x 199.4 cm) | |
| 28.5 in. | |
| 72.4 cm | |
| 58 in. | |
| 147.3 cm | |
| 52.5 in. | |
| 133.4 cm | |
| NEMA 6-15 | |
| 208-230V, 60Hz | |
| Energy efficient | |
| 277 kg |
*Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models.