missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Thermo-Seal Heat Sealing Foil

Catalog No. AB0559
Change view
Click to view available options
Material:
Aluminum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
AB0559 Aluminum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB0559 Supplier Thermo Scientific™ Supplier No. AB0559
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Suitable for long-term, low-temperature storage, transportation, and high-temperature applications such as PCR

  • Can be used in waterbath thermal cyclers
  • Foil/polypropylene laminate
  • Provides very strong seal that can be peeled only with the Foil Stripper
  • Can be pierced with a single pipet tip or pierce plate
  • Excellent chemical resistance
  • Seal Integrity Range: -80°C to +120°C

Specifications

Description Thermo-Seal; foil laminate, 85x135mm
Material Aluminum
For Use With (Application) For ALPS 25 Automated Laboratory Plate Sealer
Format Sheets