missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SUN-SRi™ Luer-Slip Syringes

Catalog No. 14823262
Change view
Click to view available options
Volume (Metric):
3 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14823262 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14823262 Supplier Thermo Scientific™ Supplier No. 400 029
Get 1 Extra Pack of Vials When You Buy 2
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific Luer Syringes offer easy, accurate and reproducible injections for a variety of volumes.

 

  • Chemically resistant, inert polypropylene and polyethylene
  • No rubber plunger seals or silicone lubricants that may cause sample contamination
  • Bulk packaged, non sterile-no paper wrap waste

 

Specifications

Material Plastic
Product Type Luer-slip Syringe
Disposable Single-use
Graduations 0.1 mL
Volume (Metric) 3 mL
Needle Point Style Centered
Connection Type Leur Slip
Sterility Non-sterile