Learn More
Thermo Scientific™ SUN-SRi™ Glass Dissolution Test Vessels
Catalog No. 22037015
Change view
Click to view available options
Material:
Borosilicate Glass
Catalog No. Material
22037015 Borosilicate Glass
1 options
Catalog No. 22037015 Supplier Thermo Scientific™ Supplier No. 100 002
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Precision formed with flat flange and hemispherical bottom
  • Manufactured from Type 1, Class A borosilicate glass Precision formed with flat flange and hemispherical bottom Glass vessels are serialized Graduations in 50mL increments, 400mL to 1000mL
Compliance
Meet all specifications of USP XXIV, Monograph 711
Specifications
Color Clear
Description Ungraduated
Material Borosilicate Glass