Learn More
Thermo Scientific™ SUN-SRi™ Dissolution Cannulae
Catalog No. 14823277
Change view
Click to view available options
For Use With:
500 mL, 7 1/2 in. Needle
900 mL, 4 3/8 in. Needle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
14823277 900 mL, 4 3/8 in. Needle
14823278 500 mL, 7 1/2 in. Needle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14823277 Supplier Thermo Scientific™ Supplier No. 401 350
Angled for easy access to samples
  • Type 316 stainless steel
  • Luer-LOK™ connection for easy attachment
  • Tip sleeve allows secure filter attachment
Order Info
All plastic syringes sold separately
Specifications
Type Cannula
For Use With 900 mL, 4 3/8 in. Needle