missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sorvall™ Stratos Microplate Rotor

Catalog No. 75003048
Change view
Click to view available options
Capacity:
2 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
75003048 2 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 75003048 Supplier Thermo Scientific™ Supplier No. 75003048

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2 x 2 microplate capacity

For use with Thermo Scientific Sorvall Stratos Centrifuges.

Specifications

Type Microtiter Plate Rotor
Capacity 2 x 1 mL
Max. RCF 1,370 x g
Max. Speed 3,000 rpm
Radius (Metric) 1.36 cm
For Use With (Equipment) Thermo Scientific™ Sorvall™ Stratus Centrifuge
Certifications/Compliance Biocontainment approved