missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sorvall ST4F Plus Centrifuge Series
GREENER_CHOICE

Catalog No. p-7214197
Change view
Click to view available options
Refrigerated:
No
Yes
Product Line:
IVD Certified
Electrical Requirements:
120 V
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Refrigerated Product Line Electrical Requirements
75009981 No IVD Certified 120 V
75009984 Yes IVD Certified 120 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 75009981 Supplier Thermo Scientific™ Supplier No. 75009981

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific Sorvall ST4F Plus Centrifuge Series

The ST4F/ST4FR Plus series floor standing centrifuges come standard with an easy-to-use keypad and high contrast LCD display. This high-capacity and versatile unit offers consistent performance and is ideal for routine applications. Quickly and securely swap between 13 unique rotor options with the tool-less Auto-Lock rotor exchange. This compact floor standing unit is on casters and can easily be moved for cleaning or storage.

  • Easy to Use:

    • Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance.
    • Modern industrial design supports continuous improvement in rotor loading ergonomics, user safety and current needed certifications
    • Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches.

    Thermo Scientific™ Auto-Lock™ III Rotor System:

    • Secure locking system allows easy push-button installation and exchange of rotors.
    • System saves time when switching between applications.
    • Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life.

    Thermo Scientific™ ClickSeal™ Bucket Sealing System:

    • Biocontainment solution seals with a snap; eliminates screw caps and complicated clips.
    • System has glove-friendly, one-handed open/close capability.
    • System is certified by CAMR™ in Porton Down, UK.

    Thermo Scientific™ SMARTSpin™ Technology:

    • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option:
    • Technology optimizes safety and improves separations.

    Space-Saving Design:

    • Compact dimensions optimize work area and ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning.

    Energy Efficient:

    • System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.

Specifications

Includes Rotor needs to be purchased separately
Product Line IVD Certified
Controller Type Microprocessor
Certifications/Compliance EN/UL/IEC 61010-1, EN/UL/IEC 61010-2-020 and 61010-2-101, EN/UL/IEC 61326-1, EN/UL/IEC 61326-2-6, FCC part 15
Capacity 4 x 1000 mL with TX-1000 Rotor
Max. Speed 15,200 rpm (Microliter 30 x 2 rotor)
Refrigerated No
Max. RCF 25,830 x g (Microliter 30 x 2 Rotor)
Rotor Included No
Safety Features SMARTSpin Imbalance detection, finger-pinch prevention, crash-proof construction
Dimensions (HWD) 32.8 x 22.3 x 27.2 in. (83.5 x 56.6 x 69 cm)
Drive System Direct, Brushless Induction Low Profile
Program Storage Up to 100 programs via push button and high contrast LCD interface
Profile (Acceleration/Braking) 9 Accel/10 Braking
Type General Purpose Centrifuge
Electrical Requirements 120 V
Display High Contrast LCD, Push Button
Ventilated Yes
Show More Show Less