Learn More
Thermo Scientific™ Sorvall™ Lid for Primo™ Round Buckets
Catalog No. 75007598
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific Sorvall Swing Bucket
Catalog No. For Use With (Equipment)
75007598 Thermo Scientific Sorvall Swing Bucket
1 options
Catalog No. 75007598 Supplier Thermo Scientific™ Supplier No. 75007598
Provides biocontainment protection
Lids fit Round Buckets (75-007-555) in 4-Place Swinging Bucket Rotor (75-007-591). Sold individually; order correct quantity for rotor.
Specifications
For Use With (Equipment) Thermo Scientific Sorvall Swing Bucket