Learn More
Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Tube Adapters for Round Rotor Buckets
For use with four-place rotors 75006445 and 75006434 when equipped with round bucket 75006441
Supplier:  Thermo Scientific™ 75008137
 View more versions of this product
Catalog No. 75008137
Only null left
Description
Description
Adapters sold individually; order correct quantity to equip your rotor.
Specifications
Chemical Identifiers
LH-4000W and LH-4000 rotor with round bucket
Specifications
LH-4000W and LH-4000 rotor with round bucket
Olive Brown
15 mL
Product Suggestions
Product Suggestions