Learn More
Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Tube Adapters for Round Rotor Buckets
For use with four-place rotors 75006445 and 75006434 when equipped with round bucket 75006441
Supplier: Thermo Scientific™ 75008136
Description
Adapters sold individually; order correct quantity to equip your rotor.
Chemical Identifiers
| LH-4000W and LH-4000 rotor with round bucket |
Specifications
| LH-4000W and LH-4000 rotor with round bucket | |
| White | |
| 10/15 mL |